Learn More
Medchemexpress LLC Sulfo Cy7 tyramide | 99.1% | 802.01 | C43H51N3O8S2 | 10 MG
SDP
Catalog No. 5000297574
Encompass_Preferred
Sulfo Cy7 tyramide is a near-infrared fluorescent tyramide reagent (Ex = 740 nm, Em = 770 nm) used as the reporter substrate for horseradish peroxidase (HRP)-catalyzed deposition in tyramide signal amplification (TSA). It is supplied as a solid with demonstrated high purity and is intended for fluorescence-based detection, including multiplex immunohistochemistry.
  • Near-infrared excitation/emission: 740/770 nm.
  • High purity (LCMS: 99.13%).
  • Compatible with HRP-based tyramide signal amplification and multiplex IHC.
  • Soluble in DMSO at 12.5 mg/mL (requires ultrasonic).
  • Store protected from light at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-97574 Supplier Medchemexpress LLC Supplier No. HYD287310MG
Sulfo Cy7 tyramide is a near-infrared fluorescent tyramide reagent (Ex = 740 nm, Em = 770 nm) used as the reporter substrate for horseradish peroxidase (HRP)-catalyzed deposition in tyramide signal amplification (TSA). It is supplied as a solid with demonstrated high purity and is intended for fluorescence-based detection, including multiplex immunohistochemistry.
  • Near-infrared excitation/emission: 740/770 nm.
  • High purity (LCMS: 99.13%).
  • Compatible with HRP-based tyramide signal amplification and multiplex IHC.
  • Soluble in DMSO at 12.5 mg/mL (requires ultrasonic).
  • Store protected from light at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).