Learn More
Medchemexpress LLC Sulfo-Cyanine3-alkyne | 1010386-62-5 | 98.1% | 100 MG
SDP
Catalog No. 5000411052
Encompass_Preferred
CY3-YNE (Sulfo-Cyanine3-alkyne) is a dye for labeling soluble proteins, peptides, and oligonucleotides/DNA. It is a click chemistry reagent containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Dye for labeling soluble proteins, peptides, and oligonucleotides/DNA
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-004-11052 Supplier Medchemexpress LLC Supplier No. HYD0818100MG
CY3-YNE (Sulfo-Cyanine3-alkyne) is a dye for labeling soluble proteins, peptides, and oligonucleotides/DNA. It is a click chemistry reagent containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Dye for labeling soluble proteins, peptides, and oligonucleotides/DNA
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups