Learn More
Medchemexpress LLC Sulfo-cyanine3 azide (potassium) | 95.3% | 736.94 | 1 MG
SDP
Catalog No. 5000432009
Encompass_Preferred
Sulfo-cyanine3 azide potassium is an azide cyanine dye and fluorescent probe (Ex/Em=548/563 nm). It can be used for synthesis of fused tricyclic heterocycles, producing immunostimulatory agents. It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Azide cyanine dye and fluorescent probe (Ex/Em=548/563 nm).
  • Used for synthesis of fused tricyclic heterocycles, producing immunostimulatory agents.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-004-32009 Supplier Medchemexpress LLC Supplier No. HYD1274C1MG
Sulfo-cyanine3 azide potassium is an azide cyanine dye and fluorescent probe (Ex/Em=548/563 nm). It can be used for synthesis of fused tricyclic heterocycles, producing immunostimulatory agents. It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Azide cyanine dye and fluorescent probe (Ex/Em=548/563 nm).
  • Used for synthesis of fused tricyclic heterocycles, producing immunostimulatory agents.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.