Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-Cyanine5-alkyne | 1345823-20-2 | 95.29% | 1 ML
SDP

Catalog No. 5000410939
Encompass_Preferred

CY5-YNE (Sulfo-Cyanine5-alkyne) is a reactive dye designed for labeling amino-groups in peptides, proteins, and oligonucleotides. This click chemistry reagent features an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.

  • Excitation: 650 nm
  • Emission: 680 nm
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Appearance: solid
  • Color: grayish brown to reddish brown

Catalog No. 50-004-10939 Supplier Medchemexpress LLC Supplier No. HYD082010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CY5-YNE (Sulfo-Cyanine5-alkyne) is a reactive dye designed for labeling amino-groups in peptides, proteins, and oligonucleotides. This click chemistry reagent features an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.

  • Excitation: 650 nm
  • Emission: 680 nm
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Appearance: solid
  • Color: grayish brown to reddish brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.