Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-cyanine5-alkyne | 1345823-20-2 | 95.3% | 693.87 g·mol⁻¹ | C36H43N3O7S2 | 10 MG
SDP

Catalog No. 5000297140
Encompass_Preferred

CY5-YNE (sulfo-Cyanine5-alkyne) is a clickable fluorescent labeling reagent containing an alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used to covalently label peptides, proteins, and oligonucleotides for fluorescence detection and imaging.

  • Alkyne functional group enables click chemistry conjugation.
  • Excitation 645 nm, emission 670 nm for Cy5-compatible detection.
  • Molecular weight 693.87 g·mol⁻¹ and formula C36H43N3O7S2.
  • Provided as solid or 10 mM solution in DMSO for flexible use.
  • Reported purity ≈95.3%; store protected from light.

Catalog No. 50-002-97140 Supplier Medchemexpress LLC Supplier No. HYD082010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CY5-YNE (sulfo-Cyanine5-alkyne) is a clickable fluorescent labeling reagent containing an alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used to covalently label peptides, proteins, and oligonucleotides for fluorescence detection and imaging.

  • Alkyne functional group enables click chemistry conjugation.
  • Excitation 645 nm, emission 670 nm for Cy5-compatible detection.
  • Molecular weight 693.87 g·mol⁻¹ and formula C36H43N3O7S2.
  • Provided as solid or 10 mM solution in DMSO for flexible use.
  • Reported purity ≈95.3%; store protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.