Learn More
Medchemexpress LLC Sulfo-Cyanine5-azide | 1621101-43-6 | 98.1% | 5 MG
SDP
Catalog No. 5000410973
Encompass_Preferred
CY5-N3 (Sulfo-Cyanine5-azide) is a Cy5-azide fluorescent dye (ex/em: 646/662 nm). It is cell membrane permeable and can be used for live cell imaging and in cell imaging by Click reaction.
  • Fluorescent dye (ex/em: 646/662 nm)
  • Cell membrane permeable
  • Can be used for live cell imaging
  • Can be used in cell imaging by Click reaction
Catalog No. 50-004-10973 Supplier Medchemexpress LLC Supplier No. HYD08325MG
CY5-N3 (Sulfo-Cyanine5-azide) is a Cy5-azide fluorescent dye (ex/em: 646/662 nm). It is cell membrane permeable and can be used for live cell imaging and in cell imaging by Click reaction.
  • Fluorescent dye (ex/em: 646/662 nm)
  • Cell membrane permeable
  • Can be used for live cell imaging
  • Can be used in cell imaging by Click reaction