Learn More
Lumiprobe Sulfo-Cyanine5 azide, 5 mg
Catalog No. NC1882675
Encompass_Preferred
Water-soluble Cyanine5 dye azide for Click chemistry. Bright and photostable dye with red fluorescence. The reagent has high water solubility. Due to high solubility and hydrophilicity, the dye can be used for labeling biomolecules in native, purely aqueous conditions.
Catalog No. NC1882675 Supplier Lumiprobe Supplier No. B33305MG
Water-soluble Cyanine5 dye azide for Click chemistry. Bright and photostable dye with red fluorescence. The reagent has high water solubility. Due to high solubility and hydrophilicity, the dye can be used for labeling biomolecules in native, purely aqueous conditions.