Learn More
Medchemexpress LLC Sulfo-cyanine5 DBCO | 981.27 | C53H57KN4O8S2 | 10 MG
SDP
Catalog No. 5000297500
Encompass_Preferred
Sulfo-Cyanine5 DBCO is a water-soluble near-infrared fluorescent dye bearing a dibenzocyclooctyne (DBCO) reactive group for copper-free click chemistry. It is intended for bioorthogonal labeling of azide-containing biomolecules and fluorescence imaging in aqueous media.
  • Near-infrared fluorescent dye (Ex/Em 646/670 nm).
  • DBCO reactive handle enables strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Water-soluble sulfonated cyanine core for labeling in aqueous buffers.
  • Suitable for conjugation to proteins, peptides, and nucleic acids.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-97500 Supplier Medchemexpress LLC Supplier No. HYD288010MG
Sulfo-Cyanine5 DBCO is a water-soluble near-infrared fluorescent dye bearing a dibenzocyclooctyne (DBCO) reactive group for copper-free click chemistry. It is intended for bioorthogonal labeling of azide-containing biomolecules and fluorescence imaging in aqueous media.
  • Near-infrared fluorescent dye (Ex/Em 646/670 nm).
  • DBCO reactive handle enables strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Water-soluble sulfonated cyanine core for labeling in aqueous buffers.
  • Suitable for conjugation to proteins, peptides, and nucleic acids.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).