Learn More
Lumiprobe Sulfo-Cyanine5 tetrazine, 1 mg
Catalog No. NC1604436
Encompass_Preferred
Sulfo-Cyanine5 tetrazine is a fluorophore derivative bearing a tetrazine group for the TCO-ligation based labeling. This reagent possesses good aqueous solubility and stability in biological environments.
Catalog No. NC1604436 Supplier Lumiprobe Supplier No. 133E0
Sulfo-Cyanine5 tetrazine is a fluorophore derivative bearing a tetrazine group for the TCO-ligation based labeling. This reagent possesses good aqueous solubility and stability in biological environments.