Learn More
Lumiprobe Sulfo-Cyanine7.5 NHS ester, 5 mg
Catalog No. NC1518298
Encompass_Preferred
Sulfo-Cyanine7.5 is a near infrared water soluble and hydrophilic dye for the NIR imaging applications.
Catalog No. NC1518298 Supplier Lumiprobe Supplier No. 26320
Sulfo-Cyanine7.5 is a near infrared water soluble and hydrophilic dye for the NIR imaging applications.