Learn More
Lumiprobe Sulfo-Cyanine7 azide, 1 mg
Catalog No. NC1328296
Encompass_Preferred
A water-soluble NIR dye azide for Click Chemistry. Sulfo-Cyanine7 is a cyanine dye, an analog of Cy7 with improved photophysical properties. This dye is a perfect choice for NIR imaging studies in live organisms.
Catalog No. NC1328296 Supplier Lumiprobe Supplier No. A5330
A water-soluble NIR dye azide for Click Chemistry. Sulfo-Cyanine7 is a cyanine dye, an analog of Cy7 with improved photophysical properties. This dye is a perfect choice for NIR imaging studies in live organisms.