Learn More
Medchemexpress LLC Sulfo-cyanine7-n3 | 97.6% | 763.95 | 1 ML
SDP
Catalog No. 5000428266
Encompass_Preferred
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble NIR dye azide for Click Chemistry. It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. It can also undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble NIR dye azide.
  • Suitable for Click Chemistry.
  • Contains an azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-004-28266 Supplier Medchemexpress LLC Supplier No. HYD105310MM/1ML
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble NIR dye azide for Click Chemistry. It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. It can also undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble NIR dye azide.
  • Suitable for Click Chemistry.
  • Contains an azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.