Learn More
Medchemexpress LLC Sulfo-cyanine7-n3 | 97.6% | 763.95 g/mol | C38H47N6O7S2 | 10 MG
SDP
Catalog No. 5000297171
Encompass_Preferred
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble near-infrared (NIR) azide dye designed for bioconjugation via click chemistry. The azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), allowing efficient fluorescent labeling of biomolecules.
  • Water-soluble NIR azide dye for click chemistry.
  • Enables CuAAC and SPAAC conjugation reactions.
  • Supplied as a green to dark green solid; also available as a DMSO solution.
  • High purity (~97.6%) suitable for labeling applications.
  • Stable when stored as a solid at 4°C and protected from light.
Catalog No. 50-002-97171 Supplier Medchemexpress LLC Supplier No. HYD105310MG
CY7-N3 (Sulfo-Cyanine7-N3) is a water-soluble near-infrared (NIR) azide dye designed for bioconjugation via click chemistry. The azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), allowing efficient fluorescent labeling of biomolecules.
  • Water-soluble NIR azide dye for click chemistry.
  • Enables CuAAC and SPAAC conjugation reactions.
  • Supplied as a green to dark green solid; also available as a DMSO solution.
  • High purity (~97.6%) suitable for labeling applications.
  • Stable when stored as a solid at 4°C and protected from light.