Learn More
Medchemexpress LLC Sulfo-cyanine7 NHS ester (TEA) | 95.8% | 907.15 g/mol | C47H62N4O10S2 | 10 MG
SDP
Catalog No. 5000297845
Encompass_Preferred
Sulfo-Cyanine7 NHS ester (TEA) is a sulfonated near-infrared fluorescent dye supplied as an NHS-activated ester (triethylammonium salt) for covalent labeling of primary amines on proteins and peptides. It enables strong NIR fluorescence for in vitro labeling, imaging, and assay development, and is supplied as a stable solid with recommended cold, dry storage.
  • Amine-reactive NHS ester for stable covalent conjugation.
  • Near-infrared fluorescence suitable for imaging and detection.
  • Water-soluble sulfonated dye for improved solubility.
  • Supplied as a solid; store sealed at 4°C and protect from light.
  • High purity appropriate for labeling applications.
Catalog No. 50-002-97845 Supplier Medchemexpress LLC Supplier No. HYD1352A10MG
Sulfo-Cyanine7 NHS ester (TEA) is a sulfonated near-infrared fluorescent dye supplied as an NHS-activated ester (triethylammonium salt) for covalent labeling of primary amines on proteins and peptides. It enables strong NIR fluorescence for in vitro labeling, imaging, and assay development, and is supplied as a stable solid with recommended cold, dry storage.
  • Amine-reactive NHS ester for stable covalent conjugation.
  • Near-infrared fluorescence suitable for imaging and detection.
  • Water-soluble sulfonated dye for improved solubility.
  • Supplied as a solid; store sealed at 4°C and protect from light.
  • High purity appropriate for labeling applications.