Learn More
Cayman Chemical Sulfo-CynIn 5 5 5mg
SDP
Catalog No. 502915180
Encompass_Preferred
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with peptides and proteins for detection by various fluorescent-based applications; has been used in cancer-targeted nanovesicles encapsulating perfluorooctyl bromide (PFOB) to identify their localization in mice; ex/em = 675/694 nm, respectively
Catalog No. 50-291-5180 Supplier Cayman Chemical Supplier No. 373695 MG
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with peptides and proteins for detection by various fluorescent-based applications; has been used in cancer-targeted nanovesicles encapsulating perfluorooctyl bromide (PFOB) to identify their localization in mice; ex/em = 675/694 nm, respectively