Learn More
Medchemexpress LLC Sulfo DBCO-amine | 2028284-70-8 | 99.0% | 50 MG
SDP
Catalog No. 5000341303
Encompass_Preferred
Sulfo DBCO-amine is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. This product is intended for research use only.
  • Designed for use in the synthesis of PROTACs.
  • Contains a DBCO group for SPAAC reactions with azide-containing molecules.
  • Classified as an alkyl chain-based linker.
  • Intended for research use only.
Catalog No. 50-003-41303 Supplier Medchemexpress LLC Supplier No. HY12705650MG
Sulfo DBCO-amine is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. This product is intended for research use only.
  • Designed for use in the synthesis of PROTACs.
  • Contains a DBCO group for SPAAC reactions with azide-containing molecules.
  • Classified as an alkyl chain-based linker.
  • Intended for research use only.