Learn More
Medchemexpress LLC Sulfo DBCO-amine | 2028284-70-8 | 99.00% | 10 MG
SDP
Catalog No. 5000342481
Encompass_Preferred
Sulfo DBCO-amine is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Utilizes intracellular ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-42481 Supplier Medchemexpress LLC Supplier No. HY12705610MG
Sulfo DBCO-amine is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Utilizes intracellular ubiquitin-proteasome system to degrade target proteins