Learn More
eMolecules​ Sulfo DBCO-Maleimide | Broadpharm | 2028281-86-7 | MFCD30458041 | 578.600 | C28H26N4O8S | 0.000 | OS(=O)(=O)C(CNC(=O)CCN1C(=O)C=CC1=O)C(=O)NCCC(=O)N1Cc2ccccc2C#Cc2ccccc12 | 25mg | 340377125
Supplier:  eMolecules​ 340377125
Encompass_Preferred
Sulfo DBCO-Maleimide | Broadpharm | 2028281-86-7 | MFCD30458041 | 578.600 | C28H26N4O8S | 0.000 | OS(=O)(=O)C(CNC(=O)CCN1C(=O)C=CC1=O)C(=O)NCCC(=O)N1Cc2ccccc2C#Cc2ccccc12 | 25mg | 340377125
Catalog No. NC1694523