Learn More
Broadpharm Sulfo DBCO-UBQ-1, 1mg Purity: 95%, Formula: C47H46N9O9S
Catalog No. NC3496333
Encompass
Sulfo DBCO-UBQ-1 serves as a click chemistry reagent uniting UBQ-1, which is a dark quencher with a polyaromatic-azo backbone that exhibits no native emission, with a sulfo group for improved hydrophilicity and DBCO for undergoing copper-free click chemistry.
Catalog No. NC3496333 Supplier Broadpharm Supplier No. BP418981MG
Sulfo DBCO-UBQ-1 serves as a click chemistry reagent uniting UBQ-1, which is a dark quencher with a polyaromatic-azo backbone that exhibits no native emission, with a sulfo group for improved hydrophilicity and DBCO for undergoing copper-free click chemistry.