Learn More
Medchemexpress LLC Sulfo-EGS 5mg | 167410-92-6 | 616.48 g/mol | C18H20N2O18S2 | 5 MG
SDP
Catalog No. 5000308351
Encompass_Preferred
Sulfo-EGS (ethylene glycol bis(sulfosuccinimidyl succinate)) is a water-soluble, homobifunctional, amine-reactive crosslinker used in biochemical research to covalently link proximal amine-containing residues and probe protein-protein or protein-peptide interactions. It is commonly used to map spatial relationships within protein complexes and to stabilize transient interactions for analysis.
  • Water-soluble crosslinker suitable for aqueous reactions.
  • Homobifunctional and targets primary amines.
  • Enables mapping of spatial proximity in protein complexes.
  • Supplied at research-scale quantity for laboratory use.
  • Typical purity approximately 90% as supplied.
Catalog No. 50-003-08351 Supplier Medchemexpress LLC Supplier No. HY1170805MG
Sulfo-EGS (ethylene glycol bis(sulfosuccinimidyl succinate)) is a water-soluble, homobifunctional, amine-reactive crosslinker used in biochemical research to covalently link proximal amine-containing residues and probe protein-protein or protein-peptide interactions. It is commonly used to map spatial relationships within protein complexes and to stabilize transient interactions for analysis.
  • Water-soluble crosslinker suitable for aqueous reactions.
  • Homobifunctional and targets primary amines.
  • Enables mapping of spatial proximity in protein complexes.
  • Supplied at research-scale quantity for laboratory use.
  • Typical purity approximately 90% as supplied.