Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfo-Egs (Ethylene Glycol Bis(Sulfosuccinimidyl Succinate))

Catalog No. 80321950MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-321-950MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-321-950MG Supplier Sigma Aldrich Supplier No. 80321950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

EGS is a water-insoluble, homobifunctional N-hydroxysuccinimide ester (NHS ester) and Sulfo-EGS is its water-soluble analog. The spacer arm contains two cleavable ester sites that may be broken with hydroxylamine, which yields two fragments with terminal amide bonds and the release of ethylene glycol. These reagents are often used for conjugating radiolabeled ligands to cell surface receptors. The water insoluble reagent EGS does not possess a charged group, is lipophilic and, therefore, membrane-permeable and useful for intracellular and intramembrane protein conjugation. EGS is first dissolved in DMSO or DMF and added to the aqueous reaction mixture at a final solvent concentration of 10-20%, to minimize detrimental affects to the protein.

Specifications

CAS 98892-75-2
Molecular Formula C62H111N3O31
Linear Formula C62H111N3O31
Quantity 100 mg
Molecular Weight (g/mol) 1394.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.