Learn More
Bioworld Sulfo-EMCS, 50 mg
Catalog No. 50199267
Encompass_Preferred
Sulfo EMCS, [N-(E-malemidocaproyloxy)-sulfosuccinimide]Primary amine reactiveSulfhydryl reactiveWater solubleReferences:Kitagawa, T., et al. (1981) Chem. Pharm. Bull. 29, 1130. 2.) Keller, O. and Rudinger, J., Helv. Chem. Acta, 58, 531.M.W. = 410.35
Catalog No. 50-199-267 Supplier Bioworld Supplier No. 218100201
Sulfo EMCS, [N-(E-malemidocaproyloxy)-sulfosuccinimide]Primary amine reactiveSulfhydryl reactiveWater solubleReferences:Kitagawa, T., et al. (1981) Chem. Pharm. Bull. 29, 1130. 2.) Keller, O. and Rudinger, J., Helv. Chem. Acta, 58, 531.M.W. = 410.35