Learn More
Medchemexpress LLC Sulfo-LC-SPDP sodium | 169751-10-4 | 527.57 | C18H22N3NaO8S3 | 10 MG
SDP
Catalog No. 5000203739
Encompass_Preferred
Sulfo-LC-SPDP sodium is a thiol-cleavable, heterobifunctional linker used to conjugate payloads to antibodies via a disulfide bond. It provides a water-soluble long-chain spacer and a pyridinyldithiol reactive group to enable payload release under reducing conditions. Intended for research use only.
  • cleavable disulfide linker for controlled payload release
  • heterobifunctional reactivity for amine and thiol conjugation
  • water-soluble sodium salt for improved aqueous handling
  • long-chain spacer reduces steric hindrance at the conjugation site
  • suitable for antibody-drug conjugate synthesis and bioconjugation workflows
  • molecular weight 527.57 and formula C18H22N3NaO8S3
Catalog No. 50-002-03739 Supplier Medchemexpress LLC Supplier No. HY126495A10MG
Sulfo-LC-SPDP sodium is a thiol-cleavable, heterobifunctional linker used to conjugate payloads to antibodies via a disulfide bond. It provides a water-soluble long-chain spacer and a pyridinyldithiol reactive group to enable payload release under reducing conditions. Intended for research use only.
  • cleavable disulfide linker for controlled payload release
  • heterobifunctional reactivity for amine and thiol conjugation
  • water-soluble sodium salt for improved aqueous handling
  • long-chain spacer reduces steric hindrance at the conjugation site
  • suitable for antibody-drug conjugate synthesis and bioconjugation workflows
  • molecular weight 527.57 and formula C18H22N3NaO8S3