Learn More
Medchemexpress LLC Sulfo-LC-SPDP (sodium) | 169751-10-4 | MFCD00083162 | >90.0% | 527.57 | C18H22N3NaO8S3 | 25 MG
SDP
Catalog No. 5000201882
Encompass_Preferred
Sulfo-LC-SPDP (sodium) is a water-soluble, cleavable heterobifunctional crosslinker that introduces a reducible disulfide linkage between amine and sulfhydryl groups. It is used as an ADC linker and for protein conjugation in research.
  • Water-soluble, cleavable heterobifunctional linker.
  • Introduces a reducible disulfide bond for controlled payload release.
  • Contains an NHS ester for reaction with primary amines.
  • Contains a pyridyldithiol group for reaction with thiols.
  • Provided as the sodium salt for improved aqueous solubility.
  • Available in small-mass quantities suitable for research-scale conjugation.
Catalog No. 50-002-01882 Supplier Medchemexpress LLC Supplier No. HY126495A25MG
Sulfo-LC-SPDP (sodium) is a water-soluble, cleavable heterobifunctional crosslinker that introduces a reducible disulfide linkage between amine and sulfhydryl groups. It is used as an ADC linker and for protein conjugation in research.
  • Water-soluble, cleavable heterobifunctional linker.
  • Introduces a reducible disulfide bond for controlled payload release.
  • Contains an NHS ester for reaction with primary amines.
  • Contains a pyridyldithiol group for reaction with thiols.
  • Provided as the sodium salt for improved aqueous solubility.
  • Available in small-mass quantities suitable for research-scale conjugation.