Learn More
Bioworld Sulfo-MBS, 100 mg
Catalog No. 50199269
Encompass_Preferred
m-Maleimidobenzoyl-N-hydroxysulfosuccinimide ester, HeterobifunctionalUseful for coupling proteins, enzymes to antibodies, toxins to antibodies, and haptens to carrier proteins.Category: Amine- and Sulfhydryl- ReactiveReactive Groups: sulfo-NHS-ester and maleimideReactive Toward: amino and sulfhydryl groupsWater Soluble: YesMembrane Permeable: NoCleavability: Non-cleavableM.W. = 416.30
Catalog No. 50-199-269 Supplier Bioworld Supplier No. 218100221
m-Maleimidobenzoyl-N-hydroxysulfosuccinimide ester, HeterobifunctionalUseful for coupling proteins, enzymes to antibodies, toxins to antibodies, and haptens to carrier proteins.Category: Amine- and Sulfhydryl- ReactiveReactive Groups: sulfo-NHS-ester and maleimideReactive Toward: amino and sulfhydryl groupsWater Soluble: YesMembrane Permeable: NoCleavability: Non-cleavableM.W. = 416.30