Learn More
Bioworld Sulfo-MPS, 50 mg
Catalog No. 50199270
Encompass_Preferred
Sulfo MPS, Maleimidopropionic acid N-hydroxysulfosuccinimide esterSulfhydryl reactivePrimary amine reactiveWater solubleM.W. = 381.27
Catalog No. 50-199-270 Supplier Bioworld Supplier No. 218100231
Sulfo MPS, Maleimidopropionic acid N-hydroxysulfosuccinimide esterSulfhydryl reactivePrimary amine reactiveWater solubleM.W. = 381.27