Learn More
Bachem Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt
SDP
Catalog No. 501947395
Encompass_Preferred
Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt, 185332-92-7, C12H11N2NaO9S, Mr 382.28, 100mg. Sulfo-GMBS sodium salt. Sulfo-N-succinimidyl 4-maleimidobutyrate (Sulfo-GMBS) is a water-soluble, heterobifunctional crosslinking reagent for conjugation of cysteine-containing peptides to carrier proteins or surfaces.
Catalog No. 50-194-7395 Supplier Bachem Supplier No. 4077463.0100
Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt, 185332-92-7, C12H11N2NaO9S, Mr 382.28, 100mg. Sulfo-GMBS sodium salt. Sulfo-N-succinimidyl 4-maleimidobutyrate (Sulfo-GMBS) is a water-soluble, heterobifunctional crosslinking reagent for conjugation of cysteine-containing peptides to carrier proteins or surfaces.