Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt | 185332-92-7 | MFCD17215919 | 98.8% | 382.28 g/mol | C12H11N2NaO9S | 1 ML
SDP

Catalog No. 5000242413
Encompass_Preferred

Sulfo-GMBS is a water-soluble, heterobifunctional cross-linker that links primary amines to sulfhydryl groups via a non-cleavable maleimide spacer. Supplied as a ready-to-use 10 mM solution in DMSO or as a solid, it is intended for protein and peptide conjugation, immobilization, and labeling in biochemical and assay workflows.

  • Water-soluble amine-thiol cross-linker suitable for protein and peptide conjugation.
  • Provides a short, non-cleavable spacer facilitating stable linkage between functional groups.
  • Available as a 10 mM solution in DMSO or in solid form for flexible use.
  • High purity (approximately 98.8%) supports reproducible conjugation reactions.
  • Soluble in DMSO at 50 mg/mL (≈130.8 mM) for easy preparation of working solutions.
  • Recommended storage: solid at -20°C sealed; in solvent at -80°C for extended stability.

Catalog No. 50-002-42413 Supplier Medchemexpress LLC Supplier No. HY10778310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfo-GMBS is a water-soluble, heterobifunctional cross-linker that links primary amines to sulfhydryl groups via a non-cleavable maleimide spacer. Supplied as a ready-to-use 10 mM solution in DMSO or as a solid, it is intended for protein and peptide conjugation, immobilization, and labeling in biochemical and assay workflows.

  • Water-soluble amine-thiol cross-linker suitable for protein and peptide conjugation.
  • Provides a short, non-cleavable spacer facilitating stable linkage between functional groups.
  • Available as a 10 mM solution in DMSO or in solid form for flexible use.
  • High purity (approximately 98.8%) supports reproducible conjugation reactions.
  • Soluble in DMSO at 50 mg/mL (≈130.8 mM) for easy preparation of working solutions.
  • Recommended storage: solid at -20°C sealed; in solvent at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.