Learn More
Medchemexpress LLC Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt | 185332-92-7 | MFCD17215919 | 98.8% | 382.28 g/mol | C12H11N2NaO9S | 10 MG
SDP
Catalog No. 5000240901
Encompass_Preferred
Sulfo-GMBS is a water-soluble, heterobifunctional cross-linking reagent containing a sulfo-NHS ester and a maleimide. It enables selective conjugation between primary amines and sulfhydryl groups in aqueous systems, making it suitable for protein and peptide bioconjugation, surface immobilization, and analytical assay development.
  • Water-soluble heterobifunctional cross-linker for aqueous reactions.
  • Reacts with primary amines via sulfo-NHS ester chemistry.
  • Reacts with sulfhydryl groups via maleimide chemistry.
  • Suitable for protein and peptide conjugation and surface immobilization.
  • Provided in small solid quantities for laboratory-scale use.
  • High purity for consistent conjugation performance.
Catalog No. 50-002-40901 Supplier Medchemexpress LLC Supplier No. HY10778310MG
Sulfo-GMBS is a water-soluble, heterobifunctional cross-linking reagent containing a sulfo-NHS ester and a maleimide. It enables selective conjugation between primary amines and sulfhydryl groups in aqueous systems, making it suitable for protein and peptide bioconjugation, surface immobilization, and analytical assay development.
  • Water-soluble heterobifunctional cross-linker for aqueous reactions.
  • Reacts with primary amines via sulfo-NHS ester chemistry.
  • Reacts with sulfhydryl groups via maleimide chemistry.
  • Suitable for protein and peptide conjugation and surface immobilization.
  • Provided in small solid quantities for laboratory-scale use.
  • High purity for consistent conjugation performance.