Learn More
Proteochem Inc Sulfo-NHS, 106627-54-7, MFCD00043100, 500 mg
Catalog No. NC0224431
Encompass_Preferred
MF: C4 H4 N O6 S . Na, MW: 217.13, Alternative Names: Sulfo NHS; Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid; S-NHS. N-hydroxysulfosuccinimide (Sulfo-NHS) is used in conjunction with EDC-HCl (EDAC-HCl) to convert carboxyl groups to Sulfo-NHS esters which react with primary amines.
Catalog No. NC0224431 Supplier Proteochem Inc Supplier No. C1102500MG
MF: C4 H4 N O6 S . Na, MW: 217.13, Alternative Names: Sulfo NHS; Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid; S-NHS. N-hydroxysulfosuccinimide (Sulfo-NHS) is used in conjunction with EDC-HCl (EDAC-HCl) to convert carboxyl groups to Sulfo-NHS esters which react with primary amines.