Learn More
Medchemexpress LLC Sulfo-NHS-Acetate | 152305-87-8 | 98.0% | C6H7NO7S | 50 MG
Supplier: Medchemexpress LLC HY14034150MG
Sulfo-NHS-Acetate is an alkyl chain-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs consist of two ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other to the target protein.
- Alkyl chain-based PROTAC linker
- Used in the synthesis of PROTACs
- PROTACs exploit the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins