Learn More
Apexbio Technology LLC Sulfo-NHS-Biotin, 119616-38-5, MFCD00078535, 100 mg
SDP
Catalog No. 501010612
Encompass_Preferred
MF: C14H18N3NaO8S2, MW: 443.4, Solubility: 22.17mg/mL in DMSO,insoluble in EtOH, 59.9 mg/mL in H2O with ultrasonic. Sulfo-NHS-Biotin (N-Hydroxysulfosuccinimidobiotin) is an amine-reactive biotinylation reagent.
Catalog No. 50-101-0612 Supplier Apexbio Technology LLC Supplier No. A8001100
MF: C14H18N3NaO8S2, MW: 443.4, Solubility: 22.17mg/mL in DMSO,insoluble in EtOH, 59.9 mg/mL in H2O with ultrasonic. Sulfo-NHS-Biotin (N-Hydroxysulfosuccinimidobiotin) is an amine-reactive biotinylation reagent.