Learn More
Cayman Chemical Sulfo-NHS-Biotin, Purity: ≥90%, CAS: 194041-65-1, 100 mg
SDP
Catalog No. 502948560
Encompass_Preferred
An amine-reactive biotinylation reagent; commonly used in the biotin labeling of amine-containing proteins, antibodies, and other molecules
Catalog No. 50-294-8560 Supplier Cayman Chemical Supplier No. 33362100 MG
An amine-reactive biotinylation reagent; commonly used in the biotin labeling of amine-containing proteins, antibodies, and other molecules