Learn More
TARGETMOL CHEMICALS INC SULFO-NHS-BIOTIN 25MG
SDP
Catalog No. 502577253
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-NHS-Biotin (Biotin-NHS) is a water-soluble biotinylation reagent used in Biotin-Streptavidin binding reactions. Sulfo-NHS-Biotin is particularly useful for targeting cell surface proteins in Streptavidin-Biotin assays. It can be used for protein detection or immobilization when used in conjunction with streptavidin and Immobilized Streptavidin. purity: 98%
Catalog No. 50-257-7253 Supplier TARGETMOL CHEMICALS INC Supplier No. T1995225MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-NHS-Biotin (Biotin-NHS) is a water-soluble biotinylation reagent used in Biotin-Streptavidin binding reactions. Sulfo-NHS-Biotin is particularly useful for targeting cell surface proteins in Streptavidin-Biotin assays. It can be used for protein detection or immobilization when used in conjunction with streptavidin and Immobilized Streptavidin. purity: 98%