Learn More
Apexbio Technology LLC Sulfo-NHS-Biotin-Kit-10-20-kDa
SDP
Catalog No. 5000567624
Encompass_Preferred
Sulfo-NHS-Biotin Kit (10-20 kDa) is a widely used reagent in the field of biomedical research particularly in protein labeling and detection studies Originating from the synthesis of sulfosuccinimidyl esters this kit functions by targeting primary amines on proteins and peptides facilitating biotinylation through a stable amide bond Its mechanism of action is based on the formation of a covalent link between biotin and cellular components enhancing detection and purification processes The in vitro activity is typically in the nanomolar to low micromolar range ensuring precise modification without compromising protein function This kit is commonly applied in cell cultures tissue extracts and animal models serving as a crucial tool in drug screening and diagnostic assay development Experimental concentrations typically depend on the specific research design allowing for flexibility and adaptability in various experimental setups
Catalog No. 50-005-67624 Supplier Apexbio Technology LLC Supplier No. K11533
Sulfo-NHS-Biotin Kit (10-20 kDa) is a widely used reagent in the field of biomedical research particularly in protein labeling and detection studies Originating from the synthesis of sulfosuccinimidyl esters this kit functions by targeting primary amines on proteins and peptides facilitating biotinylation through a stable amide bond Its mechanism of action is based on the formation of a covalent link between biotin and cellular components enhancing detection and purification processes The in vitro activity is typically in the nanomolar to low micromolar range ensuring precise modification without compromising protein function This kit is commonly applied in cell cultures tissue extracts and animal models serving as a crucial tool in drug screening and diagnostic assay development Experimental concentrations typically depend on the specific research design allowing for flexibility and adaptability in various experimental setups