Learn More
Abcam Sulfo-NHS-Biotin, NHS-ester biotinylation reagent, 250MG
Catalog No. 502705115
Encompass_Preferred
MW 443.4 Da, Purity >85%. Membrane impermeable, primary amine reactive, short chain (13.5 angstroms) NHS-ester biotinylation reagent. Targets cell surface proteins. Forms irreversible amide bonds. Biotinylates antibodies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5115 Supplier Abcam Supplier No. AB145609250MG
MW 443.4 Da, Purity >85%. Membrane impermeable, primary amine reactive, short chain (13.5 angstroms) NHS-ester biotinylation reagent. Targets cell surface proteins. Forms irreversible amide bonds. Biotinylates antibodies.
The product is subject to the following: Abcam Restricted Use Statement