Learn More
Medchemexpress LLC Sulfo-NHS-Biotin sodium | 194041-65-1 | 95.0% | 443.43 | 25 MG
SDP
Catalog No. 5000351957
Encompass_Preferred
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer (MW 443.4 Da) that can be used to assess blood-brain barrier permeability. It serves as a passive tracer for detecting blood-brain barrier leakage in mouse models of traumatic brain injury.
  • Low-molecular-weight tracer
  • Assesses blood-brain barrier permeability
  • Used as a passive tracer for detecting blood-brain barrier leakage
  • Suitable for mouse models of traumatic brain injury
  • For research use only
Catalog No. 50-003-51957 Supplier Medchemexpress LLC Supplier No. 25MGHY136964
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer (MW 443.4 Da) that can be used to assess blood-brain barrier permeability. It serves as a passive tracer for detecting blood-brain barrier leakage in mouse models of traumatic brain injury.
  • Low-molecular-weight tracer
  • Assesses blood-brain barrier permeability
  • Used as a passive tracer for detecting blood-brain barrier leakage
  • Suitable for mouse models of traumatic brain injury
  • For research use only