Learn More
Medchemexpress LLC Sulfo-NHS-biotin (sodium) | 194041-65-1 | 95.0% | 443.43 | 50 MG
SDP
Catalog No. 5000350717
Encompass_Preferred
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer used to assess blood-brain barrier permeability. It functions as a passive tracer for detecting blood-brain barrier permeability in mouse models of traumatic brain injury. No inherent pharmacodynamic effects have been observed for this product.
  • Low-molecular-weight tracer
  • Used to assess blood-brain barrier permeability
  • Detects blood-brain barrier permeability in mouse models of traumatic brain injury
  • Serves as a passive tracer
  • No inherent pharmacodynamic effects observed
Catalog No. 50-003-50717 Supplier Medchemexpress LLC Supplier No. HY13696450MG
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer used to assess blood-brain barrier permeability. It functions as a passive tracer for detecting blood-brain barrier permeability in mouse models of traumatic brain injury. No inherent pharmacodynamic effects have been observed for this product.
  • Low-molecular-weight tracer
  • Used to assess blood-brain barrier permeability
  • Detects blood-brain barrier permeability in mouse models of traumatic brain injury
  • Serves as a passive tracer
  • No inherent pharmacodynamic effects observed