Learn More
Medchemexpress LLC Sulfo-NHS-Biotin sodium | 194041-65-1 | 95.0% | 443.43 Da | 100 MG
SDP
Catalog No. 5000350289
Encompass_Preferred
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer (443.4 Da) that is used to assess blood-brain barrier permeability. It functions as a passive tracer in animal models of traumatic brain injury, helping to measure blood-brain barrier leakage volume without exhibiting inherent pharmacodynamic effects.
  • Low-molecular-weight (443.4 Da) tracer
  • Assesses blood-brain barrier permeability
  • Functions as a passive tracer
  • No inherent pharmacodynamic effects
  • Suitable for research use
Catalog No. 50-003-50289 Supplier Medchemexpress LLC Supplier No. HY136964100MG
Sulfo-NHS-Biotin sodium is a low-molecular-weight tracer (443.4 Da) that is used to assess blood-brain barrier permeability. It functions as a passive tracer in animal models of traumatic brain injury, helping to measure blood-brain barrier leakage volume without exhibiting inherent pharmacodynamic effects.
  • Low-molecular-weight (443.4 Da) tracer
  • Assesses blood-brain barrier permeability
  • Functions as a passive tracer
  • No inherent pharmacodynamic effects
  • Suitable for research use