Learn More
Medchemexpress LLC Sulfo-NHS-biotin sodium | 194041-65-1 | MFCD00078535 | ≥90.0% | C14H18N3NaO8S2 | 10MG
SDP
Catalog No. 5000259043
Encompass_Preferred
Sulfo-NHS-Biotin sodium is a water-soluble, amine-reactive biotinylation reagent that covalently labels primary amines on proteins and other biomolecules. The sulfonated N-hydroxysuccinimide ester increases aqueous solubility, allowing efficient labeling in physiological buffers and streamlined downstream analysis for detection and affinity workflows.
  • Water-soluble amine-reactive reagent for labeling primary amines.
  • Enables efficient labeling in aqueous buffers without organic solvents.
  • Supports detection, pull-downs, and affinity purification workflows.
  • High solubility in water and DMSO for flexible formulation.
  • Stable when stored sealed and protected from moisture; follow datasheet handling.
  • Available as a solid or as a pre-made DMSO solution for convenience.
Catalog No. 50-002-59043 Supplier Medchemexpress LLC Supplier No. HY13696410MG
Sulfo-NHS-Biotin sodium is a water-soluble, amine-reactive biotinylation reagent that covalently labels primary amines on proteins and other biomolecules. The sulfonated N-hydroxysuccinimide ester increases aqueous solubility, allowing efficient labeling in physiological buffers and streamlined downstream analysis for detection and affinity workflows.
  • Water-soluble amine-reactive reagent for labeling primary amines.
  • Enables efficient labeling in aqueous buffers without organic solvents.
  • Supports detection, pull-downs, and affinity purification workflows.
  • High solubility in water and DMSO for flexible formulation.
  • Stable when stored sealed and protected from moisture; follow datasheet handling.
  • Available as a solid or as a pre-made DMSO solution for convenience.