Learn More
Apexbio Technology LLC Sulfo-NHS-LC-Biotin, 100mg. Cas: 127062-22-0 MFCD: MFCD00078534. Amine-reactive biotinylation reagent, long chain
SDP
Catalog No. 501010610
Encompass_Preferred
Intermediate length, water-soluble biotinyltation reagent used to attach biotin to primary amines 127062-22-0. MFCD00078534
Catalog No. 50-101-0610 Supplier Apexbio Technology LLC Supplier No. A8003100
Intermediate length, water-soluble biotinyltation reagent used to attach biotin to primary amines 127062-22-0. MFCD00078534