Learn More
AdipoGen Sulfo-NHS-LC-Biotin (50 mg)
SDP
Catalog No. 502054314
Encompass_Preferred
Chemical. CAS: 191671-46-2127062-22-0. Formula: C20H29N4NaO9S2. MW: 556.59. Sulfo-NHS-LC-Biotin is an amino-reactive biotinylation reagent with a long spacer arm. The 6-aminocaproic acid spacer arm provides distance between the biotin and the target ligand which helps alleviate steric hindrance, in addition it forms permanent amide bonds. The sodium sulfonate groups render this compound water-soluble and prevent it from crossing the lipid bi-layer of the cell membrane, making it a tool to label cell surface proteins. In addition, Sulfo-NHS-LC-Biotin is used to biotinylate antibodies, proteins, peptides, or amino acids by reacting with primary amines as found in the N-terminus and Lysine side chains. Optimal reaction conditions are at a pH of 7-9. Avoid amine-containing buffers, which may compete in the acylation reaction.
Catalog No. 50-205-4314 Supplier AdipoGen Supplier No. CDXB0310M050
Chemical. CAS: 191671-46-2127062-22-0. Formula: C20H29N4NaO9S2. MW: 556.59. Sulfo-NHS-LC-Biotin is an amino-reactive biotinylation reagent with a long spacer arm. The 6-aminocaproic acid spacer arm provides distance between the biotin and the target ligand which helps alleviate steric hindrance, in addition it forms permanent amide bonds. The sodium sulfonate groups render this compound water-soluble and prevent it from crossing the lipid bi-layer of the cell membrane, making it a tool to label cell surface proteins. In addition, Sulfo-NHS-LC-Biotin is used to biotinylate antibodies, proteins, peptides, or amino acids by reacting with primary amines as found in the N-terminus and Lysine side chains. Optimal reaction conditions are at a pH of 7-9. Avoid amine-containing buffers, which may compete in the acylation reaction.