Learn More
Abcam Sulfo-NHS-LC-Biotin, biotinylating reagent, 10MG
Catalog No. 502705112
Encompass_Preferred
MW 556.6 Da, Purity >90%. Sulfo-NHS-LC-Biotin, biotinylating reagent. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5112 Supplier Abcam Supplier No. AB14561110MG
MW 556.6 Da, Purity >90%. Sulfo-NHS-LC-Biotin, biotinylating reagent. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement