Learn More
TARGETMOL CHEMICALS INC SULFO-NHS-LC-BIOTIN SODI 50MG
SDP
Catalog No. 502576856
Encompass_Preferred
Also available in 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-NHS-LC-Biotin sodium is an amine-reactive biotinylation reagent for antibody labeling. purity: 98%
Catalog No. 50-257-6856 Supplier TARGETMOL CHEMICALS INC Supplier No. T3934050MG
Also available in 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-NHS-LC-Biotin sodium is an amine-reactive biotinylation reagent for antibody labeling. purity: 98%