Learn More
Medchemexpress LLC Sulfo-NHS-LC-Biotin sodium | 191671-46-2 | 98.0% | 100 MG
SDP
Catalog No. 5000409713
Encompass_Preferred
Sulfo-NHS-LC-Biotin sodium is an amine-reactive ester derivative of biotin designed for assessing vascular permeability. It is particularly useful as it cannot pass the intact blood-retinal barrier, making it suitable for studying the brain and retinal vasculature.
  • Amine-reactive ester derivative of biotin
  • Cannot pass the intact blood-retinal barrier
  • Used to assess vascular permeability of brain and retinal vasculature
  • Suitable for protein biotinylation
  • Also used for cell surface protein biotinylation
Catalog No. 50-004-09713 Supplier Medchemexpress LLC Supplier No. HYD0799100MG
Sulfo-NHS-LC-Biotin sodium is an amine-reactive ester derivative of biotin designed for assessing vascular permeability. It is particularly useful as it cannot pass the intact blood-retinal barrier, making it suitable for studying the brain and retinal vasculature.
  • Amine-reactive ester derivative of biotin
  • Cannot pass the intact blood-retinal barrier
  • Used to assess vascular permeability of brain and retinal vasculature
  • Suitable for protein biotinylation
  • Also used for cell surface protein biotinylation