Learn More
Medchemexpress LLC Sulfo-NHS-LC-Biotin sodium | 191671-46-2 | 98.01% | 1 ML
SDP
Catalog No. 5000410371
Encompass_Preferred
Sulfo-NHS-LC-Biotin sodium is an amine-reactive ester derivative of biotin that cannot pass the intact blood-retinal barrier. It can be used to assess vascular permeability of the brain and retinal vasculature, making it suitable for research applications.
  • Amine-reactive ester derivative of biotin
  • Cannot pass the intact blood-retinal barrier
  • Assess vascular permeability of the brain and retinal vasculature
  • Suitable for research use only
Catalog No. 50-004-10371 Supplier Medchemexpress LLC Supplier No. HYD079910MM/1ML
Sulfo-NHS-LC-Biotin sodium is an amine-reactive ester derivative of biotin that cannot pass the intact blood-retinal barrier. It can be used to assess vascular permeability of the brain and retinal vasculature, making it suitable for research applications.
  • Amine-reactive ester derivative of biotin
  • Cannot pass the intact blood-retinal barrier
  • Assess vascular permeability of the brain and retinal vasculature
  • Suitable for research use only