Learn More
Apexbio Technology LLC Sulfo-NHS-LC-D-Desthiobiotin 5X1mg
SDP
Catalog No. 5000092152
Encompass_Preferred
Sulfo-NHS-LC-D-Desthiobiotin is a sulfonated amine-reactive labeling reagent targeting primary amines on proteins or peptides It is designed to covalently label cell-surface proteins by reacting with lysine residues or protein N-termini thereby enabling selective modification of these proteins Sulfo-NHS-LC-D-Desthiobiotin exerts its biological activity primarily through covalent reaction with primary amines via its activated Sulfo-NHS ester Due to its membrane-impermeable negatively charged sulfonate group it selectively labels cell-surface proteins without entering intact cells Based on these chemical properties Sulfo-NHS-LC-D-Desthiobiotin holds research potential in affinity purification biotin-streptavidin interaction assays selective enrichment and analytical workflows involving reversible labeling of surface proteins
Catalog No. 50-000-92152 Supplier Apexbio Technology LLC Supplier No. A80185
Sulfo-NHS-LC-D-Desthiobiotin is a sulfonated amine-reactive labeling reagent targeting primary amines on proteins or peptides It is designed to covalently label cell-surface proteins by reacting with lysine residues or protein N-termini thereby enabling selective modification of these proteins Sulfo-NHS-LC-D-Desthiobiotin exerts its biological activity primarily through covalent reaction with primary amines via its activated Sulfo-NHS ester Due to its membrane-impermeable negatively charged sulfonate group it selectively labels cell-surface proteins without entering intact cells Based on these chemical properties Sulfo-NHS-LC-D-Desthiobiotin holds research potential in affinity purification biotin-streptavidin interaction assays selective enrichment and analytical workflows involving reversible labeling of surface proteins