Learn More
Bioworld Sulfo-NHS LC SPDP, 100 mg
Catalog No. 50199271
Encompass_Preferred
Sulfo NHS LC SPDP, Sulfosuccinimidyl 6-[3-(2-pyridyldithio) propionamido] HexanoateWater solubleSpacer arm SPDPAntibody-toxin conjugationReferences: 1.) Carlsson, J., et al. (1978) Biochem. J. 173, 723-737.M.W. = 527.6
Catalog No. 50-199-271 Supplier Bioworld Supplier No. 218100241
Sulfo NHS LC SPDP, Sulfosuccinimidyl 6-[3-(2-pyridyldithio) propionamido] HexanoateWater solubleSpacer arm SPDPAntibody-toxin conjugationReferences: 1.) Carlsson, J., et al. (1978) Biochem. J. 173, 723-737.M.W. = 527.6