Learn More
Apexbio Technology LLC Sulfo-NHS-SS-Biotin, 325143-98-4, MFCD00078533, 100 mg
SDP
Catalog No. 501010608
Encompass_Preferred
MF: C19H27N4NaO9S4, MW: 606.7, Solubility: 30.335mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Sulfo-NHS-SS-biotin(sulfosuccinimidyl-20(biotinamido)ethyl-1,3-dithiopropionate) is a long-chain cleavable amine-reactive biotinylation reagent.
Catalog No. 50-101-0608 Supplier Apexbio Technology LLC Supplier No. A8005100
MF: C19H27N4NaO9S4, MW: 606.7, Solubility: 30.335mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Sulfo-NHS-SS-biotin(sulfosuccinimidyl-20(biotinamido)ethyl-1,3-dithiopropionate) is a long-chain cleavable amine-reactive biotinylation reagent.