Learn More
Medchemexpress LLC HY-111496 10mg Medchemexpress, Sulfo-NHS-SS-Biotin (sodium) CAS:325143-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11149610MG
Encompass_Preferred
Medchemexpress, HY-111496 10mg Sulfo-NHS-SS-Biotin (sodium) CAS:325143-98-4 Sulfo-NHS-SS-biotin is a long-chain cleavable and cell-impermeant amine-reactive biotinylation reagent. Sulfo-NHS-SS-biotin can be used for the labeling and purifying of cell-surface protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0793